Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma Mouse anti-Human, Alexa Fluor™ 750, Clone: 43-11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP231044AF750

Catalog No. NP231044A70


Add to cart

Description

Description

IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

IFN-gamma
43-11
Western Blot, ELISA
Affinity Purified, Conjugated
IFNG
Monoclonal Antibody to Human IFN-g (ELISA Capture)-Azide Free
Immunogen affinity purified
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Primary
3458.0
Human
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody recognizes both the recombinant and the natural form of human interferon-gamma (IFN-gamma) (1). This antibody does not react with human IL-4, IFN-alpha or acid inactivated IFN-gamma, and it shows only a minimal reactivity with heatinactivated IFN-g (<0.05%) (1).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit