Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-gamma R2 Antibody (MOB47), Alexa Fluor™ 532, Novus Biologicals™
Hamster Monoclonal Antibody
Supplier: Novus Biologicals FAB773X
Description
IFN-gamma R2 Monoclonal antibody specifically detects IFN-gamma R2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOFSpecifications
IFN-gamma R2 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Hamster | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3460 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Flow Cytometry, Neutralization, CyTOF | |
MOB47 | |
Western Blot, Flow Cytometry, Neutralization, CyTOF-ready | |
AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFN-gamma-R2, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1 | |
Recombinant mouse IFN-gamma R2, Extracellular domain | |
0.5 mL | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction