Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma R2 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI1C2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271008JF549

Catalog No. N271008J549


Add to cart

Description

Description

IFN-gamma R2 Monoclonal antibody specifically detects IFN-gamma R2 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

IFN-gamma R2
OTI1C2
Immunohistochemistry
Affinity Purified, Conjugated
IFNGR2
Full length recombinant protein of human IFN-gamma R2 (NP_005525) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFN-gamma-R2, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1
Mouse
IgG2b
0.1 mL
Primary
3460.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit