Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma, Rabbit anti-Human,Baboon, Clone: D9D10, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$265.00 - $566.50

Specifications

Antigen IFN-gamma
Clone D9D10
Concentration 1 mg/mL
Dilution Western Blot, Flow Cytometry, ELISA, Block/Neutralize
Applications Western Blot, Flow Cytometry, ELISA, Blocking Assay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB014721
SDP
View Documents
Novus Biologicals
NBP2810440.025MG
0.025 mg
Each for $265.00
Add to Cart
 
NB014722
SDP
View Documents
Novus Biologicals
NBP2810440.2MG
0.2 mg
Each for $566.50
Add to Cart
 
Description

Description

IFN-gamma Monoclonal specifically detects IFN-gamma in Human, Baboon samples. It is validated for Western Blot, Flow Cytometry, ELISA, Block/Neutralize.
Specifications

Specifications

IFN-gamma
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Blocking Assay
Unconjugated
Rabbit
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
PBS
3458
FPLC-purified natural human IFN-gamma. Refer to the original article [PMID: 3114127]
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
D9D10
Western Blot, Flow Cytometry, ELISA, Block/Neutralize
Monoclonal
Purified
RUO
Human, Baboon
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
IFNG
IgG κ
Protein A purified
Binds human IFN-gamma, a dimerized soluble cytokine that is the only member of the type II class of interferons.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit