Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-gamma Rabbit anti-Human, Clone: 506, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | IFN-gamma |
---|---|
Clone | 506 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for ELISA.Specifications
IFN-gamma | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
Recombinant Human IFN-gamma Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
506 | |
ELISA | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
0.2 um filtered solution in PBS | |
3458 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title