Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-omega/IFNW1 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen IFN-omega/IFNW1
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093650
View Documents
Novus Biologicals
NBP303585100UL
100 μg Each for $320.71
Add to cart
 
NB093651
View Documents
Novus Biologicals
NBP30358520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

IFN-omega/IFNW1 Polyclonal antibody specifically detects IFN-omega/IFNW1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

IFN-omega/IFNW1
Unconjugated
Rabbit
Cell Cycle and Replication
PBS with 50% glycerol, pH7.3.
3467
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry, Western Blot
Purified
RUO
Mouse, Rat
IFN-omega 1, interferon omega-1, Interferon alpha-II-1, interferon omega-1, interferon, omega 1
Recombinant fusion protein containing a sequence corresponding to amino acids 22-195 of human IFN-omega/IFNW1 (NP_002168.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit