Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNA16 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550684

Catalog No. PA550684


Add to cart

Description

Description

The antibody detects endogenous levels of total IFNA16 protein.

IFNA16 is produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications

Specifications

IFNA16
Polyclonal
Unconjugated
IFNA16
Liquid
IFNA16
Synthetic peptide corresponding to residues near the C terminal of human interferon, alpha 16
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
2.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P05015
IFN-alphaO
Rabbit
IgG
100 μL
-20°C
Primary
3449
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit