Learn More
IFNA16 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS6330R
Catalog No. 50-198-7203
Add to cart
Description
Description
IFNA16 is produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
Specifications
IFNA16
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IFNA16
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
3449
IFNA16
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
P05015
IFNA16; IFN-alpha-16; IFN-alpha-N-protein; IFN-alphaO; interferon alpha 16; Interferon alpha-16; Interferon alpha-WA; interferon, alpha 16
KLH conjugated synthetic peptide derived from human IFNA16.
Protein A
RUO
Antibody
Polyclonal
Human