Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNA5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549021

Catalog No. PA549021


Add to cart

Description

Description

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications

Specifications

IFNA5
Polyclonal
PBS with 0.09% sodium azide
P01569
Interferon alpha-5, IFN-alpha-5, Interferon alpha-61, Interferon alpha-G, LeIF G, IFNA5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3442
Western Blot
Unconjugated
IFNA5
Liquid
IFNA5
KLH conjugated synthetic peptide between 163-189 amino acids from the C-terminal region of human IFNA5.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit