Sino Biological™ IFNB1 Recombinant Mouse Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain of mouse IFNB1 (Met 1-Asn 182) fused to Fc region of human IgG1 at C-terminus

Manufacturer: Sino Biological™ 50708M02H5

 View more versions of this product

Catalog No. 50-708-M02H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 46.7kd
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Source Mouse
    Structural Form Truncated, Recombinant
    Endotoxin Concentration <1 EU/μg
    Gene Symbol IFNB1
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Expression System Mammalian
    Product Type IFNB1 Recombinant Mouse Protein
    For Use With (Application) Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 15977
    Common Name Interferon Beta 1, Fibroblast
    Protein Family Other Proteins

    Expressed from DNA sequence encoding extracellular domain of mouse IFNB1 (Met 1-Asn 182) fused to Fc region of human IgG1 at C-terminus

    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit