Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNG Mouse anti-Bovine, Clone: 2E6, Abnova™

Mouse monoclonal antibody raised against bovine IFNG.

Manufacturer:  Abnova Corporation MAB12441

Catalog No. 89-934-344


Add to cart

Description

Description

Specifications

Specifications

IFNG
2E6
Mouse monoclonal antibody raised against bovine IFNG.
Lyophilized from PBS, pH 7.2 (0.09% sodium azide)
IFNG
Native purified bovine IFNG.
Antigen Affinity Purification
RUO
Primary
281237
ELISA, Immunoprecipitation, Western Blot
Unconjugated
ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Lyophilized
Mouse
IgG1
1 mg
Store at 4°C on dry atmosphere. After reconstitution with deionized water, store at −20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Bovine
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit