Learn More
Abnova™ Ifng (Mouse) ELISA Kit
Sandwich enzyme immunoassay for the quantitative measurement of mouse Ifng.
Manufacturer:  Abnova™ KA4813
Catalog No. 89-948-397
Add to cart
Description
Description
  • Used for the in vitro quantitative determination of Mouse IFNγ in Mouse serum, Mouse plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant Mouse IFNγ
Specifications
Specifications
P01580
3.9 to 250 pg/mL
Ifng (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Ifng.
15978
ELISA Kit
100 μL
Mouse
Colorimetric
Biotin
Quantification
Mouse IFNγ microtiter plate
For research use only (RUO)
Store the kit at 4°C.
Buffered Solution, Cell Lysates, Culture Supernatant, Plasma, Serum