Sino Biological™ IFNW1 Recombinant Human Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human IFNW1 (Cys 24-Ser 195) fused to Fc region of human IgG1 at N-terminus

Manufacturer: Sino Biological™ 10353H01H5

 View more versions of this product

Catalog No. 10-353-H01H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Structural Form Full Length, Recombinant
    Molecular Weight 46.6kd
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Source Human
    Endotoxin Concentration <1 EU/μg
    Common Name Interferon, omega-1
    Gene Symbol IFNW1
    Protein Tag hIgG1-Fc Tag
    Validated Application Assay Standard
    Protein Family Cytokines and Receptors
    Expression System Mammalian
    Product Type IFNW1 Recombinant Human Protein
    For Use With (Application) Performing cell based assays
    Format Lyophilized

    Expressed from DNA sequence encoding human IFNW1 (Cys 24-Ser 195) fused to Fc region of human IgG1 at N-terminus

    • Measured in antiviral assays using WISH human amnion cells infected with vesicular stomatitis virus (VSV)
    • EC50 for effect is 0.1-0.5ng/mL
    • Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8 mM KH2
  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit