Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ifosfamide, Acros Organics™

Click to view available options
Quantity:
1g
5g
3778-73-2
C7H15Cl2N2O2P
261.09
HOMGKSMUEGBAAB-UHFFFAOYSA-N
ifosfamide,isophosphamide,iphosphamide,isofosfamide,ifosfamid,mitoxana,naxamide,iphosphamid,isoendoxan,ifex
3690
CHEBI:5864
N,3-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine
C1CN(P(=O)(OC1)NCCCl)CCCl
3778-73-2
C7H15Cl2N2O2P
261.09
HOMGKSMUEGBAAB-UHFFFAOYSA-N
ifosfamide,isophosphamide,iphosphamide,isofosfamide,ifosfamid,mitoxana,naxamide,iphosphamid,isoendoxan,ifex
3690
CHEBI:5864
N,3-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine
C1CN(P(=O)(OC1)NCCCl)CCCl
Melting Point 39.0°C to 42.0°C
Infrared Spectrum Authentic
Quantity 1g
Water 0.5% max.
Formula Weight 261.09
Percent Purity ≥97.5% (ELSD) (HPLC)
Chemical Name or Material Ifosfamide

GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/

GHS Signal Word: Danger

EINECSNumber : 223-237-3