Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFRD2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548833
Description
Specifications
IFRD2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q12894 | |
IFRD2 | |
KLH conjugated synthetic peptide between 478-506 amino acids from the C-terminal region of human IFRD2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
IFRD2 | |
Interferon-related developmental regulator 2, Protein SKMC15, IFRD2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7866 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction