Learn More
IFRD2 purified MaxPab mouse Mouse anti-Human, Polyclonal, Abnova™
Mouse Polyclonal Antibody
Manufacturer:  Abnova Corporation H00007866B01P
Catalog No. 89-032-202
Add to cart
Description
Description
Mouse polyclonal antibody raised against a full-length human IFRD2 protein.
  • High specificity
  • High reactivity
Specifications
Specifications
IFRD2 purified MaxPab mouse
Unconjugated
In 1x PBS, pH 7.4
Liquid
IFRD2
IFRD2 (NP_006755.3, 1 a.a. ~ 442 a.a) full-length human protein.
RUO
Primary
7866
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human IFRD2 protein.
NM_006764.3
FLJ40446|IFNRP|SKMc15|SM15
Mouse
50 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)