Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IFT140 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541317
Description
Peptide sequence: VLRWSPSGNC LLSGDRLGVL LLWRLDQRGR VQGTPLLKHE YGKHLTHCIF Sequence homology: Cow: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
IFT140 contains 9 TPR repeats and 5 WD repeats. The function of the IFT140 protein remains unknown.
Specifications
IFT140 | |
Polyclonal | |
Unconjugated | |
IFT140 | |
AI661311; c305C8.4; c380F5.1; gs114; Ift140; intraflagellar transport 140; intraflagellar transport 140 homolog; intraflagellar transport protein 140 homolog; KIAA0590; LOW QUALITY PROTEIN: intraflagellar transport protein 140 homolog; mKIAA0590; MZSDS; SRTD9; Tce5; T-complex expressed gene 5; WD and tetratricopeptide repeats 2; WD and tetratricopeptide repeats protein 2; WDTC2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9742 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96RY7 | |
IFT140 | |
Synthetic peptide directed towards the N-terminal of human IFT140 (aa 108-157). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction