Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFT88 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179538

 View more versions of this product

Catalog No. NBP179538


Add to cart

Description

Description

IFT88 Polyclonal antibody specifically detects IFT88 in Human, Mouse, Rat, Canine, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

IFT88
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
IFT88
Synthetic peptide directed towards the middle region of human IFT88The immunogen for this antibody is IFT88. Peptide sequence EIREQRIKSGRDGSGGSRGKREGSASGDSGQNYSASSKGERLSARLRALP.
91 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Predicted Homology Based On Immunogen Sequence: Rat: 86%; Rabbit: 86%; Pig: 85%; Horse: 85%; Mouse: 85%; Guinea pig: 85%.
Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
NP_006522
D13S1056ETPR repeat protein 10, DAF19, hTg737, intraflagellar transport 88 homolog (Chlamydomonas), intraflagellar transport protein 88 homolog, MGC26259, polaris homolog, probe hTg737 (polycystic kidney disease, autosomal recessive), Recessive polycystic kidney disease protein Tg737 homolog, tetratricopeptide repeat domain 10, Tetratricopeptide repeat protein 10, TG737Tg737, TTC10
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
8100
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit