Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig gamma-4 Chain C Region, Rabbit anti-Human, DyLight 550, Clone: IGHG4/2042R, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279850R
Description
Ig gamma-4 Chain C Region Monoclonal antibody specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Ig gamma-4 Chain C Region | |
IGHG4/2042R | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
IGHG4 | |
Recombinant full-length human Ig gamma-4 Chain C Region protein | |
Protein A purified | |
RUO | |
Primary | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker) | |
Rabbit | |
IgG | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title