Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig Goat anti-Rat, APC, Polyclonal, Southern Biotech™
Goat Polyclonal Secondary Antibody
$294.00 - $493.00
Specifications
Antigen | Ig |
---|---|
Concentration | 0.5mg/mL |
Applications | Flow Cytometry, ELISA |
Classification | Polyclonal |
Conjugate | APC |
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
Ig | |
Flow Cytometry, ELISA | |
APC | |
RUO | |
Supplied in PBS/NaN3 and a stabilizing agent | |
IgG | |
Affinity Purified |
0.5mg/mL | |
Polyclonal | |
Goat | |
Rat | |
Rat IgG and IgM | |
Secondary | |
Store at 2-8°C; Protect from light; Do not freeze |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title