Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig kappa light chain Mouse anti-Human, Clone: Kap-56, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14290
Description
Mouse monoclonal antibody raised against native human Ig kappa light chain.
- High specificity
- High reactivity
Specifications
Ig kappa light chain | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-2 ug/106 cells in 0.1 mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
P01834 | |
IGKC | |
Human lymphocytes stimulated. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot | |
Kap-56 | |
Mouse monoclonal antibody raised against native human Ig kappa light chain. | |
In 10 mM PBS. | |
HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809 | |
Mouse | |
Protein A/G purification | |
RUO | |
3514 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction