Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig kappa light chain Rabbit anti-Human, Clone: RM126, Abnova™

Rabbit monoclonal antibody raised against native human Ig kappa light chain.

Manufacturer:  Abnova Corporation MAB12809

Catalog No. 89-946-942


Add to cart

Specifications

Specifications

Ig kappa light chain
RM126
Rabbit monoclonal antibody raised against native human Ig kappa light chain.
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
HCAK1|Km|MGC111575|MGC62011|MGC72072|MGC88770|MGC88771|MGC88809
Rabbit
IgG
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody reacts to the kappa light chain of human immunoglobulins. No cross reactivity with the lambda light chain, mouse IgG, rat IgG, or goat IgG.
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed)
Unconjugated
ELISA (0.05-0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
IGKC
Native purified IgG from human plasma.
Protein A affinity purification
RUO
Primary
3514
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit