Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig, κ light chain Rat anti-Mouse, Brilliant Violet 510, Clone: 187.1, BD Optibuild
Rat Monoclonal Antibody
Manufacturer: BD Biosciences 742834
Description
The 187.1 monoclonal antibody specifically binds to kappa light chains of mouse immunoglobulins. The 187.1 antibody does not react with mouseλ1 orλ2 immunoglobulin lights chains or mouse immunoglobulin heavy chains.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.Specifications
Ig, κ light chain | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Ig kappa chain C region, IGKC, Igk-C, Ig, κ | |
Mouse IgG2b κ secreted by MPC-11 plasmacytoma | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV510 under optimal conditions that minimize unconjugated dye and antibody. | |
RUO | |
Primary | |
Murine |
187.1 | |
Brilliant Violet 510 | |
Affinity Purified | |
Rat | |
Rat SD, also known as Sprague-Dawley (outbred) IgG1, κ | |
50μg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title