Learn More
IgA Fc fragment Sheep anti-Porcine, Polyclonal, Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP269213
Catalog No. NBP2-69213
Add to cart
Description
Description
IgA Fc fragment Polyclonal antibody specifically detects IgA Fc fragment in Porcine samples. It is validated for Radioimmune Assays, Immunoelectrophoresis.
Specifications
Specifications
IgA Fc fragment
LYOPH
Radioimmunodiffusion, Immunoelectrophoresis
Antiserum, Lyophilized
Highly purified normal IgA isolated from pooled swine serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
Secondary
Precipitating polyclonal Sheep antiSerum to Swine IgA, Fc specific.
Porcine
Immunoelectrophoresis, Immunofluorescence, Immunohistochemistry (Frozen), Radioimmune Assays
Unconjugated
Whole antiserum with No Preservative
Sheep
Whole Antiserum
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute the lyophilized antiserum by adding 1 ml sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).