Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgA Human, HRP, Clone: , Isotype Control, Southern Biotech™
Human Isotype Control
Supplier: Southern Biotechnology Associates 0155L05
Description
SouthernBiotech isotype controls and purified immunoglobulins are useful as negative controls and standards in a variety of immunoassays.
Specifications
ELISA | |
HRP | |
Human | |
RUO | |
Store at 2-8°C |
Lot specific | |
Supplied in a stock solution in 50% glycerol/50% PBS, pH 7.4; No preservative added | |
1.0 mL | |
Primary | |
IgA |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction