Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA, kappa, BB515, Clone: M18-254, Isotype Control, BD

Manufacturer:  BD Biosciences 565095

Catalog No. BDB565095


Add to cart

Description

Description

Specifications

Specifications

IgA, κ
M18-254
Brilliant Blue 515
Affinity purified
IgA, κ
4°C, protect from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide
Murine
50μg
Secondary
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit