Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Equine IgA Secondary Antibody (K129.2G5), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP268461
Description
IgA Monoclonal specifically detects IgA in Equine samples. It is validated for ELISA.Specifications
IgA | |
Monoclonal | |
1 mg/ml | |
ELISA 1:20000 - 1:200000 | |
Immunoglobulin A | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Immunology | |
3493.0 | |
Store at 4°C. | |
IgG1 |
ELISA | |
K129.2G5 | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
IGHA1 | |
Equine IgA | |
0.1 mg | |
Secondary | |
Equine | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction