Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgA Mouse anti-Human, Clone: rHISA43, Novus Biologicals™
Mouse Monoclonal Antibody
$277.00
Specifications
Antigen | IgA |
---|---|
Clone | rHISA43 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122316
|
Novus Biologicals
NBP30742120UG |
20 μg |
Each of 1 for $277.00
|
|
|||||
Description
IgA Monoclonal specifically detects IgA in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
IgA | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Immunology | |
10 mM PBS with 0.05% BSA | |
3493 | |
IgG1 κ | |
Protein A or G purified |
rHISA43 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Immunoglobulin A | |
Recombinant full-length human IGHA1 and IGHA2 proteins | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title