Learn More
Athens Research And Technology IncSupplier Diversity Partner IGA1 HU MYELOMA PLASMA 1 MG DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 16160907011M1MG
Immunoglobulin A1, Human Myeloma Plasma: 1 MG; Frozen in 10 mM phosphate, pH 7.4, 150 mM NaCl and 0.05% NaN3; MW 160,000; EC=1.32; Purity: > =95% by SDS-PAGE. Native; Prepared and purified from human myeloma plasma in the Athens Research & Tech lab.
Catalog No. 50-489-764