Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGBP1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI4F6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271009AF647

Catalog No. N271009A647


Add to cart

Description

Description

IGBP1 Monoclonal antibody specifically detects IGBP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

IGBP1
OTI4F6
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
IGBP1
Full length recombinant protein of human IGBP1 (NP_001542) produced in HEK293T cells.
Immunogen affinity purified
Protein Phosphatase, Signal Transduction
Primary
3476.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
ALPHA-4, B cell signal transduction molecule alpha 4, bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ), B-cell signal transduction molecule alpha 4, CD79a-binding protein 1, IBP1, immunoglobulin (CD79A) binding protein 1, immunoglobulin-binding protein 1, Protein alpha-4, protein phosphatase 2A, regulatory subunit alpha-4, Renal carcinoma antigen NY-REN-16
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit