Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD[b] Mouse anti-Mouse, Brilliant Violet 510, Clone: 217-170, BD Optibuild™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 742384

Catalog No. BDB742384


Add to cart

Description

Description

The 217-170 monoclonal antibody specifically recognizes mouse IgD of Igh-C[b] /haplotype (eg, C57BL/6, C57BL/10, SJL). It does not react with IgD of Igh-C[a] /or related haplotypes (eg, BALB/c, AKR, CBA, C3H/He, C58, DBA/1, DBA/2, NZB, PL, RIII), or Igh-C[e] /haplotype (eg, A). 217-170 antibody is effective for detection of cell-surface Ig by immunofluorescent staining with flow cytometric analysis. It has not been shown to stimulate B-cell proliferation.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications

Specifications

IgD[b]
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Igh-5b, Ighd, immunoglobulin heavy constant delta
Not reported
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV510 under optimal conditions that minimize unconjugated dye and antibody.
RUO
Primary
Murine
217-170
Brilliant Violet 510
Affinity Purified
Murine
Mouse BALB/c IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit