Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD[b] Mouse anti-Mouse, Brilliant Violet 605, Clone: 217-170, BD Optibuild™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 742385

Catalog No. BDB742385


Add to cart

Description

Description

The 217-170 monoclonal antibody specifically recognizes mouse IgD of Igh-C[b] /haplotype (eg, C57BL/6, C57BL/10, SJL). It does not react with IgD of Igh-C[a] /or related haplotypes (eg, BALB/c, AKR, CBA, C3H/He, C58, DBA/1, DBA/2, NZB, PL, RIII), or Igh-C[e] /haplotype (eg, A). 217-170 antibody is effective for detection of cell-surface Ig by immunofluorescent staining with flow cytometric analysis. It has not been shown to stimulate B-cell proliferation.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
Specifications

Specifications

IgD[b]
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Igh-5b, Ighd, immunoglobulin heavy constant delta
Not reported
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV605 under optimal conditions that minimize unconjugated dye and antibody.
RUO
Primary
Murine
217-170
Brilliant Violet 605
Affinity Purified
Murine
Mouse BALB/c IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit