Learn More
IgD Mouse anti-Human, Alexa Fluor 405, Clone: IA6-2, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9857V100UG
Catalog No. FB9857V100
Add to cart
This item is not returnable. View return policy
Description
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
IgD
IA6-2
Flow Cytometry 0.25-1 μg/106 cells
Purified, Supernatant, Conjugated
Mouse
IgG
100 μg
Primary
3495
Human
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Human IgD,
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human IgD in Flow Cytometry.