Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Antibody (11-26c.2a) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280784
Description
IgD Monoclonal specifically detects IgD in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
IgD | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10-1:1000, CyTOF-ready | |
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
11-26c.2a | |
Unconjugated | |
PBS | |
IGHD | |
This antibody was generated by immunizing host with mouse IgD | |
0.5 mg | |
B Cell Development and Differentiation Markers, Immunology | |
3495 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction