Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGDCC3 Alexa Fluor 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8559U100UG
This item is not returnable.
View return policy
Description
IGDCC3 Monoclonal specifically detects IGDCC3 in Human samples. It is validated for Flow Cytometry.Specifications
IGDCC3 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
HsT18880, immunoglobulin superfamily DCC subclass member 3, immunoglobulin superfamily, DCC subclass, member 3, PUNC, Putative neuronal cell adhesion moleculePUNCHsT18880 | |
Human embryonic kidney cell line HEK293-derived recombinant human IGDCC3 Met1-Gly640 Accession # Q81VU1 | |
RUO | |
9543 | |
Human | |
IgG1 |
Flow Cytometry | |
920038 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q81VU1 | |
Mouse | |
100 μg | |
Primary | |
Detects human IGDCC3 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction