Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgE Fc Porcine anti-Human, HRP, Polyclonal, Secondary Antibody, Novus Biologicals
Porcine Polyclonal Antibody
Manufacturer: Novus Biologicals NBP269116
Description
IgE Fc Polyclonal antibody specifically detects IgE Fc in Human samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Dot Blot.Specifications
IgE Fc | |
an aliquot is thawed slowly at ambient temperature and spun down again. | |
HRP | |
Lyophilized from PBS (pH 7.2) with No Preservative | |
Porcine | |
Ion exchange chromatography | |
divided into small aliquots | |
Polyclonal | |
Reconstitute by by adding 1 ml sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. |
ELISA, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
LYOPH | |
ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000 | |
Conjugated, Lyophilized | |
Purified monoclonal IgE isolated from human serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
1 mL | |
Secondary | |
Horseradish peroxidase-conjugated IgG fraction of polyclonal Swine antiSerum to Human IgE, Fc specific. | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title