Learn More
Enzo Life SciencesSupplier Diversity Partner IgE (non-immune) (h) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner BPDDIAHE101
Alternative name: Immunoglobulin E. Purity: ≥98% (SDS-PAGE). Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.15M sodium chloride. Source: Purified human IgE with kappa light chains produced in vitro from a monoclonal hybridoma. Application: ICC. FUNC
Catalog No. 50-659-115