Learn More
Igepal(R) CA-630, Spectrum™
Click to view available options
Quantity:
25 mL
100 mL
500 mL
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
9002-93-1
C16H26O2
250.38
JYCQQPHGFMYQCF-UHFFFAOYSA-N
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
9002-93-1
C16H26O2
250.38
JYCQQPHGFMYQCF-UHFFFAOYSA-N
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
I1112, 9002-93-1, (C2H4O)nC14H22O, n=9-10
Spectrum™ Igepal(R) CA-630 is a non-denaturing detergent. It is derived from octylphenol which serves as the hydrophobic part. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
CAS Max % 1
Packaging Steel Can
Quantity 20 L