Learn More
Drg International Inc Igf-1 600 Elisa
Supplier:  Drg International Inc EIA4140
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of Insulin-like Growth Factor 1 (IGF-1) in serum. Insulin-like growth factor I (IGF-1) is a polypeptide of 70 amino acids (7650 daltons), and is one of a number of related insulin-like growth factors present in the circulation. A single basal IGF-1 determination is useful in the assessment of short stature in children and in nutritional support studies of acutely ill patients. For the diagnosis of acromegaly, a single IGF-1 determination is considered more reliable than a random GH determination. The DRG IGF-1 600 ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. Patient samples, standards and controls are acidified and neutralized prior to the assay procedure. The microtiter wells are coated with a monoclonal antibody directed towards an antigenic site on the IGF-1 molecule.
Catalog No. 50-194-7604
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.