Learn More
IGF-1R (α-subunit) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ IGF-1R (α-Subunit) Ab-1, Mouse Monoclonal Antibody.
$652.00 - $841.00
Specifications
Antigen | IGF-1R (alpha-Subunit) Ab-1 |
---|---|
Clone | 24-31 |
Applications | Immunoprecipitation |
Classification | Monoclonal |
Form | Concentrated |
Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity & Availability | |||
MS641B1
|
Epredia
MS641B1 |
Biotin | 200μg/mL | 500 μL |
Each for $841.00
|
|
|||
MS641P1
|
Epredia
MS641P1 |
Unconjugated | 200μg/mL | 500 μL |
Each for $652.00
|
|
|||
MS641B0
|
Epredia
MS641B0 |
Biotin | 200μg/mL | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS641B
|
Epredia
MS641B |
Biotin | 200μg/mL | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS641P0
|
Epredia
MS641P0 |
Unconjugated | 200μg/mL | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS641P
|
Epredia
MS641P |
Unconjugated | 200μg/mL | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS641P1ABX
|
Epredia
MS641P1ABX |
Unconjugated | 1mg/mL | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS641PABX
|
Epredia
MS641PABX |
Unconjugated | 1mg/mL | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of α- and Β-subunits, which are disulfide-linked in a Β-α-α-Β configuration in the mature receptor. The α-subunit is completely extracellular, while the Β-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.Host Species: Mouse
Clone: 24-31
Isotype: IgG1
Species Reactivity: Human. Weakly reacts with Rabbit. Does not react with rat. Pig unknown.
Epitope: aa 283-440 (exon 4-6)
Immunogen: IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Molecular Weight: 125kDa
Positive Control: Placenta or Breast CA
Cellular Localization: Membrane
Recommended for:
- Western Blotting (Not suitable)
- Immunoprecipitation
- ELISA
- Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)
- Does Not Significantly Inhibit IGF-1 Binding
Note:
USA: RUO; Int'l: RUO
Specifications
IGF-1R (alpha-Subunit) Ab-1 | |
Immunoprecipitation | |
Concentrated | |
RUO | |
Human | |
IgG1 |
24-31 | |
Monoclonal | |
Mouse | |
Neuroscience | |
IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA | |
Primary |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.