Learn More
IGF-1R (α-subunit) Ab-4 Mouse Monoclonal Antibody, Epredia™
Epredia™ IGF-1R (α-Subunit) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Epredia MS644P1ABX
Description
IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of α- and Β-subunits, which are disulfide-linked in a Β-α-α-Β configuration in the mature receptor. The α-subunit is completely extracellular, while the Β-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.Host Species: Mouse
Clone: 24-60
Isotype: IgG2a/κ
Species Reactivity: Human. Weakly reacts with Rabbit and Pig (Ab-2 is better). Does not react with rat.
Epitope: aa 184-283
Immunogen: IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Molecular Weight: 125kDa
Positive Control: Placenta or Breast CA
Cellular Localization: Membrane Ab-4 is excellent for immunoprecipitation of alpha-subunit of IGF-1R.
Recommended for:
- Western Blotting (Not suitable)
- Immunoprecipitation
- Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)
- Substantially (>80%) Inhibits IGF-1 Binding
Note:
USA: RUO; Int'l: RUO
Specifications
IGF-1R (alpha-Subunit) Ab-4 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Neuroscience | |
Human | |
IgG2a κ |
Immunoprecipitation | |
24-60 | |
Unconjugated | |
IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.