Learn More
IGF-I Hamster anti-Mouse, Janelia Fluor 549, Clone: RM0099-2F54, Novus Biologicals™
Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP122455JF549
Catalog No. NBP12455549
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IGF-IMonoclonalantibody specifically detects IGF-I in Mouse samples.
Specifications
Specifications
IGF-I
RM0099-2F54
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
IGF1
Mouse recombinant protein of IGF-I
Protein G purified
Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction
Primary
3479
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C
Hamster
IgG2
0.1mL
Store at 4°C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a Hamster immunized with mouse recombinant protein of IGF-I.