Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-I/IGF-1 Rabbit anti-Human, Mouse, Rat, Rabbit, Clone: IGF1/2872R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen IGF-I/IGF-1
Clone IGF1/2872R
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Flow Cytometry, ELISA, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122236
View Documents
Novus Biologicals
NBP30738120UG
20 μg
Each for $159.82
Add to cart
 
NB122235
View Documents
Novus Biologicals
NBP30738110UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

IGF-I/IGF-1 Monoclonal antibody specifically detects IGF-I/IGF-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications

Specifications

IGF-I/IGF-1
0.2 mg/ml
Flow Cytometry, ELISA, Immunofluorescence
Purified
RUO
Human, Mouse, Rat, Rabbit
IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C
Recombinant full-length human IGF-I/IGF-1 protein
Monoclonal
Protein A or G purified
IGF1/2872R
Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Rabbit
Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction
10 mM PBS with 0.05% BSA
3479
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit