Learn More
IGF-I R/IGF1R, Mouse anti-Human, Alexa Fluor 488, Clone: 33255, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB391G100UG
Catalog No. FAB391G100
Add to cart
This item is not returnable. View return policy
Description
Description
IGF-I R/IGF1R Monoclonal specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
IGF-I R/IGF1R
33255
Flow Cytometry 0.25-1 ug/10^6 cells
P08069
CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IGF-I R/IGF1R, Glu31-Asn932, Accession # P08069
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3480
Human
Flow Cytometry
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Mouse
IgG1
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human IGF-I R/IGF1R in sandwich ELISAs and Western blots. In sandwich immunoassays, less than 0.15% cross-reactivity or interference was observed with recombinant human (rh) IGF-I, rhIGF-II, rhIL-3 R alpha , rhIL-9 R, and rhTGF-beta RII.