Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF1R (CD221) Recombinant Rabbit Monoclonal Antibody (012), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA546646

Encompass

Catalog No. PIMA546646


Add to Cart

Description

Description

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human IGF1R (CD221) (Met1-Asn932) and conjugated with PE under optimum conditions, the unreacted PE was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.

IGF1R (CD221) Recombinant Monoclonal antibody specifically detects IGF1R (CD221) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IGF1R (CD221)
Recombinant Monoclonal
100 μg/mL
PBS with 0.5% BSA and 0.09% sodium azide
P08069
IGF1R
Recombinant protein ecoding Human IGF1R (CD221) (Met1-Asn932).
100 Tests
Primary
Human
Antibody
IgG
Flow Cytometry
012
PE
IGF1R
A330103N21Rik; CD221; CD221 antigen; D930020L01; hyft; IGF 1 receptor; IGF 1R; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; Insulin like growth factor 1 receptor precursor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Rabbit
Protein A
RUO
3480
4° C, store in dark, DO NOT FREEZE!
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit