Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF1R Mouse anti-Human, Clone: JBW902, Abnova™

Mouse monoclonal antibody raised against human IGF1R

Manufacturer:  Abnova Corporation MAB12559

Catalog No. 89-934-352


Add to cart

Description

Description

This receptor binds insulin-like growth factor with a high affinity. It has tyrosine kinase activity. The insulin-like growth factor I receptor plays a critical role in transformation events. Cleavage of the precursor generates alpha and beta subunits. It is highly overexpressed in most malignant tissues where it functions as an anti-apoptotic agent by enhancing cell survival. [provided by RefSeq]

Specifications

Specifications

IGF1R
JBW902
Mouse monoclonal antibody raised against human IGF1R
In 0.1M Tris-glycine, 0.15M NaCl, pH 7.4 (0.05% sodium azide).
CD221/IGFIR/JTK13/MGC142170/MGC142172/MGC18216
Mouse
IgG2a
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Liquid
IGF1R
Recombinant GST fusion protein corresponding to amino acids 1264-1367 of human IGF1R.
Protein A purification
RUO
Primary
3480
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit