Learn More
MBL International Corporation Anti-IGF2BP3 (IMP3) (Human/Mouse) pAb (Polyclonal Antibody)
SDP
Supplier:  MBL International Corporation RN009P
Encompass_Preferred
Clonality:Polyclonal; Application:IP|RIP|WB; Conjugate:Unlabeled; Species Reactivity:Human|Mouse; Size:200 µl; Target:IGF2BP3/IMP3; IGF2BP3/IMP3 is identical to the KH domain containing protein overexpressed in cancer (KOC). The protein encoded by this gene was found to bind in vivo to IGF2 leader 3 and leader 4 mRNAs and H19 RNA. Binding of IMP3 to the 5' UTR of the IGF2 leader 3 mRNA represses translation of IGF2 during late development.Polyclonal Antibody of 200 µl targeting IGF2BP3/IMP3 for IP, RIP, WB.IGF2BP3/IMP3 Polyclonal Antibody.
Catalog No. 50-510-21
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.