Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-1 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI6C7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271011AF405

Catalog No. N271011A405


Add to cart

Description

Description

IGFBP-1 Monoclonal antibody specifically detects IGFBP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

IGFBP-1
OTI6C7
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
IGFBP1
Recombinant protein fragment corresponding to amino acids 26-259 of human IGFBP-1 (NP_000587) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP
Mouse
IgG1
0.1 mL
Primary
3484.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit