Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals IGFBP-3 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Manufacturer: Novus Biologicals NBP2794625PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IGFBP-3.Specifications
31.25 pg/mL | |
Sandwich ELISA | |
RUO | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IGFBP-3. | |
IGFBP-3 | |
Apoptosis, Biologically Active Proteins, Prostate Cancer | |
Human | |
IGFBP3 | |
1. Capture Antibody: Dilute to a working concentration of 0.5 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2 ng/mL is recommended. |
HRP | |
3486 | |
Antibody Pairs | |
31.25-2000 pg/mL | |
Storage is content dependent. | |
Rabbit | |
5 Plates | |
Sandwich ELISA | |
acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title