Learn More
IGFBP-4 Mouse, Alexa Fluor 647, Clone: IBP180, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232771AF647
Catalog No. NBP232771AH
Add to cart
Description
Description
IGFBP-4 Monoclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
IGFBP-4
IBP180
ELISA
Purified
IGFBP4
Human N-terminal IGFBP-4 fragment (NT-IGFBP-4)
Protein A purified
RUO
Primary
3487
ELISA
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
BP-4, HT29-IGFBP, IBP4insulin-like growth factor-binding protein 4, IGF-binding protein 4, IGFBP-4IBP-4, insulin-like growth factor binding protein 4
Mouse
IgG2a
0.2 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse